accessibility menu, dialog, popup

UserName

CH3ON(Z)

CAS RN 61310-53-0

IUPAC Name: (2Z)-3-ethoxyprop-2-enenitrile
Synonyms: (2Z)-3-ethoxyprop-2-enenitrile
Molecular Weight (g/mol): 97.12
Molecular Formula: C5H7NO
InChi Key: HUPVIAINOSTNBJ-HYXAFXHYSA-N
SMILES: CCO\C=C/C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results