missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-N,N-diethylaniline 97.0+%, TCI America™

Catalog No. M103925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M103925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M103925G Supplier TCI America Supplier No. M103925G
Only null left

Chemical Identifiers

CAS 1864-92-2
Molecular Formula C12H19NO
Molecular Weight (g/mol) 193.29
MDL Number MFCD00043936
InChI Key ODQSBWZDOSNPAH-UHFFFAOYSA-N
Synonym N,N-Diethyl-m-phenetidine
PubChem CID 74625
IUPAC Name 3-ethoxy-N,N-diethylaniline
SMILES CCN(CC)C1=CC(=CC=C1)OCC

Specifications

Color Yellow
Boiling Point 270°C
Quantity 25 g
Formula Weight 193.29
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Ethoxy-N,N-diethylaniline
EINECSNumber (3)-0694
TSCA Yes