accessibility menu, dialog, popup

UserName

CH3ONH3CH3C

CAS RN 1864-92-2

IUPAC Name: 3-ethoxy-N,N-diethylaniline
Synonyms: 3-ethoxy-N,N-diethylaniline
Molecular Weight (g/mol): 193.29
Molecular Formula: C12H19NO
InChi Key: ODQSBWZDOSNPAH-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(=C1)N(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results