missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-methoxybenzonitrile 98.0+%, TCI America™

Catalog No. p-7129335
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11095G 5 g
E110925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11095G Supplier TCI America Supplier No. E11095G

Chemical Identifiers

CAS 60758-86-3
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD02256160
InChI Key XTIINWPNAMHVDG-UHFFFAOYSA-N
PubChem CID 3934769
IUPAC Name 3-ethoxy-4-methoxybenzonitrile
SMILES CCOC1=C(C=CC(=C1)C#N)OC

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 177.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxy-4-methoxybenzonitrile