missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-methoxybenzoic acid, 98%

Catalog No. AAB2048903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2048903 1 g
AAB2048906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2048903 Supplier Thermo Scientific Chemicals Supplier No. B2048903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2651-55-0 | C10H11O4 | 195.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2651-55-0
Molecular Formula C10H11O4
Molecular Weight (g/mol) 195.20
MDL Number MFCD00276591
InChI Key DMSAIFTWQMXOBE-UHFFFAOYSA-M
Synonym benzoic acid,3-ethoxy-4-methoxy, maybridge1_005010, 3-ethoxy-4-methoxybenzoicacid, acmc-1cqm3
PubChem CID 2822957
IUPAC Name 3-ethoxy-4-methoxybenzoic acid
SMILES CCOC1=CC(=CC=C1OC)C([O-])=O

Specifications

Melting Point 156°C to 160°C
Quantity 1 g
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 3-Ethoxy-4-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only