missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxy-4-hydroxybenzaldehyde 98.0+%, TCI America™
$40.43 - $273.38
Chemical Identifiers
| CAS | 121-32-4 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00006944 |
| InChI Key | CBOQJANXLMLOSS-UHFFFAOYSA-N |
| Synonym | ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal |
| PubChem CID | 8467 |
| ChEBI | CHEBI:48408 |
| IUPAC Name | 3-ethoxy-4-hydroxybenzaldehyde |
| SMILES | CCOC1=CC(C=O)=CC=C1O |
Chemical Identifiers
| 121-32-4 | |
| 166.18 | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| 8467 | |
| 3-ethoxy-4-hydroxybenzaldehyde |
| C9H10O3 | |
| MFCD00006944 | |
| ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal | |
| CHEBI:48408 | |
| CCOC1=CC(C=O)=CC=C1O |
Specifications
| 121-32-4 | |
| White-Yellow | |
| MFCD00006944 | |
| ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal | |
| CCOC1=CC(C=O)=CC=C1O | |
| 166.18 | |
| CHEBI:48408 | |
| ≥98.0% (HPLC,T) | |
| 3-Ethoxy-4-hydroxybenzaldehyde |
| 78°C | |
| C9H10O3 | |
| 25 g | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| 3-ethoxy-4-hydroxybenzaldehyde | |
| 8467 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1201
RTECSNumber : CU6125000
TSCA : Yes