missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-hydroxybenzaldehyde, 98%

CAS: 121-32-4 | C9H10O3 | 166.18 g/mol

$42.01 - $285.99

Chemical Identifiers

CAS 121-32-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00006944
InChI Key CBOQJANXLMLOSS-UHFFFAOYSA-N
Synonym ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal
PubChem CID 8467
ChEBI CHEBI:48408
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
SMILES CCOC1=CC(C=O)=CC=C1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1947814
View Documents
Thermo Scientific Chemicals
A1947814
25 g
Each for $42.01
Only null left
 
AAA1947822
View Documents
Thermo Scientific Chemicals
A1947822
100 g
Each for $69.89
Only null left
 
AAA1947836
View Documents
Thermo Scientific Chemicals
A1947836
500 g
Each for $285.99
Only null left
 
Description

Description

It plays an important role as an antioxidant and is also used as a flavoring agent, in perfumes, and in organic synthesis. It holds anti-angiogenic, anti-inflammatory and antinociceptive properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role as an antioxidant and is also used as a flavoring agent, in perfumes, and in organic synthesis. It holds anti-angiogenic, anti-inflammatory and antinociceptive properties.

Solubility
Slightly soluble in water. Soluble in ethanol, ether, benzene.

Notes
Air & light sensitive. Protect from light. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

121-32-4
166.18
CBOQJANXLMLOSS-UHFFFAOYSA-N
8467
3-ethoxy-4-hydroxybenzaldehyde
C9H10O3
MFCD00006944
ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal
CHEBI:48408
CCOC1=CC(C=O)=CC=C1O

Specifications

121-32-4
285°C
C9H10O3
25 g
Air and light sensitive
ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal
CBOQJANXLMLOSS-UHFFFAOYSA-N
3-ethoxy-4-hydroxybenzaldehyde
8467
166.18
3-Ethoxy-4-hydroxybenzaldehyde
75°C to 79°C
127°C (260°F)
MFCD00006944
1073761
14,3859
Slightly soluble in water. Soluble in ethanol,ether,benzene.
CCOC1=CC(C=O)=CC=C1O
166.18
CHEBI:48408
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

EINECSNumber : 204-464-7

RTECSNumber : CU6125000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only