missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxy-4-hydroxybenzaldehyde, 97%
CAS: 121-32-4 | C9H10O3 | 166.18 g/mol
$69.24 - $249.59
Chemical Identifiers
| CAS | 121-32-4 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00006944 |
| InChI Key | CBOQJANXLMLOSS-UHFFFAOYSA-N |
| Synonym | ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal |
| PubChem CID | 8467 |
| ChEBI | CHEBI:48408 |
| IUPAC Name | 3-ethoxy-4-hydroxybenzaldehyde |
| SMILES | CCOC1=CC(C=O)=CC=C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC156031000
|
Thermo Scientific Chemicals
156031000 |
100 g | Glass bottle |
Each for $69.24
|
|
||||
|
AC156035000
|
Thermo Scientific Chemicals
156035000 |
500 g | Glass bottle |
Each for $249.59
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 121-32-4 | |
| 166.18 | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| 8467 | |
| 3-ethoxy-4-hydroxybenzaldehyde |
| C9H10O3 | |
| MFCD00006944 | |
| ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal | |
| CHEBI:48408 | |
| CCOC1=CC(C=O)=CC=C1O |
Specifications
| 121-32-4 | |
| 100.0 | |
| White | |
| 96% min. (GC) | |
| C9H10O3 | |
| MFCD00006944 | |
| 08,256 | |
| ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| 3-ethoxy-4-hydroxybenzaldehyde | |
| 8467 | |
| 166.18 | |
| Fine Crystalline Powder |
| 96.0 | |
| 76.0°C to 79.0°C | |
| Authentic | |
| Glass bottle | |
| C2H5OC6H3(OH)CHO | |
| 100 g | |
| 15,3908 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,glycerol and prpylene,glycol | |
| CCOC1=CC(C=O)=CC=C1O | |
| 166.18 | |
| CHEBI:48408 | |
| 97% | |
| 3-Ethoxy-4-hydroxybenzaldehyde, 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 204-464-7
RTECSNumber : CU6125000
TSCA : TSCA
RUO – Research Use Only