missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2,4-difluorobenzoic acid, 97%

Catalog No. p-7030916
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3181203 1 g
AAH3181206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3181203 Supplier Thermo Scientific Chemicals Supplier No. H3181203

CAS: 1017779-88-2 | C9H8F2O3 | 202.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1017779-88-2
Molecular Formula C9H8F2O3
Molecular Weight (g/mol) 202.157
MDL Number MFCD09258723
InChI Key KUOUFQUWQYELBO-UHFFFAOYSA-N
Synonym 3-ethoxy-2,4-difluorobenzoic acid, jrd
PubChem CID 46737640
IUPAC Name 3-ethoxy-2,4-difluorobenzoic acid
SMILES CCOC1=C(C=CC(=C1F)C(=O)O)F

Specifications

Melting Point 134°C to 136°C
Quantity 1 g
Formula Weight 202.15
Percent Purity 97%
Chemical Name or Material 3-Ethoxy-2,4-difluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only