missing translation for 'onlineSavingsMsg'
Learn More

3-Dodecylthiophene 98.0+%, TCI America™

Catalog No. D20161G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20161G 1 g
D20165G 5 g
D201625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D20161G Supplier TCI America Supplier No. D20161G

Chemical Identifiers

CAS 104934-52-3
Molecular Formula C16H28S
Molecular Weight (g/mol) 252.46
MDL Number MFCD00130121,MFCD31699966
InChI Key RFKWIEFTBMACPZ-UHFFFAOYSA-N
Synonym 3-laurylthiophene, thiophene, 3-dodecyl, poly, 3-n-dodecylthiophene, 3-dodecyl thiophene, 1-thien-3-yl dodecane, acmc-1boc7, 3-dodecylthiophene, ksc174a5h, bidd:gt0253
PubChem CID 2733655
IUPAC Name 3-dodecylthiophene
SMILES CCCCCCCCCCCCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 252.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Dodecylthiophene