accessibility menu, dialog, popup

UserName

H3CS

CAS RN 104934-52-3

IUPAC Name: 3-dodecylthiophene
Synonyms: 3-dodecylthiophene
Molecular Weight (g/mol): 252.46
Molecular Formula: C16H28S
InChi Key: RFKWIEFTBMACPZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CSC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)