Learn More
3-(Diphenylphosphino)propylamine, 97+%
CAS: 16605-03-1 | C15H18NP | 243.29 g/mol
Supplier: Thermo Scientific Chemicals H6067203
Description
3-(Diphenylphosphino)propylamine is used as a paharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Diphenylphosphino)propylamine is used as a paharmaceutical.
Solubility
Not miscible or difficult to mix in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
Specifications
| 3-(Diphenylphosphino)propylamine | |
| C15H18NP | |
| MFCD00233832 | |
| UN2735 | |
| Air Sensitive | |
| Not miscible or difficult to mix in water. | |
| NCCCP(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 243.29 | |
| 243.28 |
| 16605-03-1 | |
| (C6H5)2PCH2CH2CH2NH2 | |
| 1 g | |
| 2730850 | |
| 3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine | |
| DQZWMOWSTWWMPP-UHFFFAOYSA-N | |
| 3-diphenylphosphanylpropan-1-amine | |
| 280677 | |
| ≥97% |
Chemical Identifiers
| 16605-03-1 | |
| 243.29 | |
| DQZWMOWSTWWMPP-UHFFFAOYSA-N | |
| 280677 | |
| NCCCP(C1=CC=CC=C1)C1=CC=CC=C1 |
| C15H18NP | |
| MFCD00233832 | |
| 3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine | |
| 3-diphenylphosphanylpropan-1-amine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only