accessibility menu, dialog, popup

UserName

CAS RN 16605-03-1

IUPAC Name: (3-aminopropyl)diphenylphosphane
Synonyms: (3-aminopropyl)diphenylphosphane
Molecular Weight (g/mol): 243.29
Molecular Formula: C15H18NP
InChi Key: DQZWMOWSTWWMPP-UHFFFAOYSA-N
SMILES: NCCCP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)