missing translation for 'onlineSavingsMsg'
Learn More

3-(Diphenylphosphino)-1-propylamine 97.0+%, TCI America™

Catalog No. D4825200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4825200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4825200MG Supplier TCI America Supplier No. D4825200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16605-03-1
Molecular Formula C15H18NP
Molecular Weight (g/mol) 243.29
MDL Number MFCD00233832
InChI Key DQZWMOWSTWWMPP-UHFFFAOYSA-N
Synonym 3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine
PubChem CID 280677
IUPAC Name (3-aminopropyl)diphenylphosphane
SMILES NCCCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 200 mg
UN Number 2735
Formula Weight 243.29
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 3-(Diphenylphosphino)-1-propylamine