Learn More
3-Dimethylaminopropyl chloride hydrochloride, 98%
CAS: 5407-04-5 | C5H13Cl2N | 158.07 g/mol
Supplier: Thermo Scientific Chemicals A154590E
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Dimethylaminopropyl chloride hydrochloride | |
| 138°C to 144°C | |
| MFCD00012521 | |
| 3670046 | |
| 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
| CN(C)CCCCl.Cl | |
| 158.07 | |
| 158.07 |
| 4-5-5407 | |
| C5H13Cl2N | |
| 2500 g | |
| Hygroscopic | |
| LJQNMDZRCXJETK-UHFFFAOYSA-N | |
| 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride | |
| 94308 | |
| 98% |
Chemical Identifiers
| 4-5-5407 | |
| 158.07 | |
| LJQNMDZRCXJETK-UHFFFAOYSA-N | |
| 94308 | |
| CN(C)CCCCl.Cl |
| C5H13Cl2N | |
| MFCD00012521 | |
| 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
| 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 226-467-2
RTECSNumber : UI0209950
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only