missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminopropyl chloride hydrochloride, 98%

Catalog No. p-7022603
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1545922 100 g
AAA1545936 500 g
AAA154590E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1545922 Supplier Thermo Scientific Chemicals Supplier No. A1545922
Only null left

CAS: 5407-04-5 | C5H13Cl2N | 158.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4-5-5407
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.07
MDL Number MFCD00012521
InChI Key LJQNMDZRCXJETK-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride
PubChem CID 94308
IUPAC Name 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCCl.Cl

Specifications

Melting Point 138°C to 144°C
Quantity 100 g
Beilstein 3670046
Sensitivity Hygroscopic
Formula Weight 158.07
Percent Purity 98%
Chemical Name or Material 3-Dimethylaminopropyl chloride hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-467-2
RTECSNumber UI0209950
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only