missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminobenzoyl chloride hydrochloride, tech. 90%

Catalog No. p-7047268
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1843303 1 g
AAL1843306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1843303 Supplier Thermo Scientific Chemicals Supplier No. L1843303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 117500-61-5 | C9H11Cl2NO | 220.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117500-61-5
Molecular Formula C9H11Cl2NO
Molecular Weight (g/mol) 220.093
MDL Number MFCD02094023
InChI Key NUDQPATXNFWFNK-UHFFFAOYSA-N
Synonym 3-dimethylaminobenzoyl chloride hydrochloride, 3-dimethylamino benzoyl chloride hydrochloride, acmc-20an50, 3-dimethylaminobenzoyl chloride hcl, 3-dimethylamino-benzoyl chloride hydrochloride salt, 3-dimethylamino benzoyl chloride-hydrogen chloride 1/1
PubChem CID 22732309
IUPAC Name 3-(dimethylamino)benzoyl chloride;hydrochloride
SMILES CN(C)C1=CC=CC(=C1)C(=O)Cl.Cl

Specifications

Melting Point 131°C to 133°C
Quantity 1 g
UN Number UN3261
Beilstein 2803337
Sensitivity Moisture sensitive
Formula Weight 220.1
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Dimethylaminobenzoyl chloride hydrochloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only