accessibility menu, dialog, popup

UserName

CAS RN 117500-61-5

IUPAC Name: 3-(dimethylamino)benzoyl chloride hydrochloride
Synonyms: 3-(dimethylamino)benzoyl chloride hydrochloride
Molecular Weight (g/mol): 220.09
Molecular Formula: C9H11Cl2NO
InChi Key: NUDQPATXNFWFNK-UHFFFAOYSA-N
SMILES: Cl.CN(C)C1=CC=CC(=C1)C(Cl)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)