missing translation for 'onlineSavingsMsg'
Learn More

3'-Dimethylaminoacetophenone 98.0+%, TCI America™

Catalog No. p-7126733
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
D14465G 5 g
D144625G 25 g
2 options

Catalog No. D14465G

Supplier: TCI America D14465G

Only null left

Chemical Identifiers

CAS 18992-80-8
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00059343
InChI Key KYEMJVGXLJXCSM-UHFFFAOYSA-N
PubChem CID 519620
IUPAC Name 1-[3-(dimethylamino)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC=C1)N(C)C

Specifications

Melting Point 37°C
Quantity 5 g
Formula Weight 163.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3′-Dimethylaminoacetophenone