accessibility menu, dialog, popup

UserName

CAS RN 18992-80-8

IUPAC Name: 1-[3-(dimethylamino)phenyl]ethan-1-one
Synonyms: 1-[3-(dimethylamino)phenyl]ethanone
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: KYEMJVGXLJXCSM-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(=C1)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results