missing translation for 'onlineSavingsMsg'
Learn More

3'-Dimethylaminoacetophenone 98.0+%, TCI America™

Catalog No. D144625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D144625G 25 g
D14465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D144625G Supplier TCI America Supplier No. D144625G

Chemical Identifiers

CAS 18992-80-8
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00059343
InChI Key KYEMJVGXLJXCSM-UHFFFAOYSA-N
PubChem CID 519620
IUPAC Name 1-[3-(dimethylamino)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC=C1)N(C)C

Specifications

Melting Point 37°C
Quantity 25 g
Formula Weight 163.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3′-Dimethylaminoacetophenone