missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)propiophenone Hydrochloride 98.0+%, TCI America™

Catalog No. D240025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D240025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D240025G Supplier TCI America Supplier No. D240025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 879-72-1
Molecular Formula C11H16ClNO
Molecular Weight (g/mol) 213.705
MDL Number MFCD00012481
InChI Key DKNDBIIKSJWQFL-UHFFFAOYSA-N
Synonym 3-dimethylaminopropiophenone hydrochloride, 3-dimethylamino-1-phenylpropan-1-one hydrochloride, 3-dimethylamino propiophenone hydrochloride, beta-dimethylaminopropiophenone hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride, nsc 89, dimethylaminoethylphenylketone hydrochloride, propiophenone, 3-dimethylamino-, hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride 1:1, wln: 1n1&2vr &gh
PubChem CID 92844
IUPAC Name 3-(dimethylamino)-1-phenylpropan-1-one;hydrochloride
SMILES CN(C)CCC(=O)C1=CC=CC=C1.Cl

Specifications

Melting Point 153°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 213.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)propiophenone Hydrochloride
RTECSNumber UH0600000
TSCA Yes