accessibility menu, dialog, popup

UserName

HClH3CNCH3O

CAS RN 879-72-1

IUPAC Name: hydrogen 3-(dimethylamino)-1-phenylpropan-1-one chloride
Synonyms: hydrogen 3-(dimethylamino)-1-phenylpropan-1-one chloride
Molecular Weight (g/mol): 213.71
Molecular Formula: C11H16ClNO
InChi Key: DKNDBIIKSJWQFL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results