missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)propionic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. D26905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26905G 5 g
D269025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26905G Supplier TCI America Supplier No. D26905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14788-12-6
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD02093464
InChI Key JTNKXYWGZCNBCH-UHFFFAOYSA-N
Synonym N,N-Dimethyl-beta-alanine Hydrochloride
PubChem CID 11586327
IUPAC Name 3-(dimethylamino)propanoic acid;hydrochloride
SMILES CN(C)CCC(=O)O.Cl

Specifications

Melting Point 189°C
Color White
Quantity 5 g
Formula Weight 153.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)propionic Acid Hydrochloride