accessibility menu, dialog, popup

UserName

H3CNCH3OOHHCl

CAS RN 14788-12-6

IUPAC Name: 3-(dimethylamino)propanoic acid hydrochloride
Synonyms: 3-(dimethylamino)propanoic acid hydrochloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: JTNKXYWGZCNBCH-UHFFFAOYSA-N
SMILES: Cl.CN(C)CCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results