missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)benzoic Acid 98.0+%, TCI America™

Catalog No. D0646500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0646500G 500 g
D064625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0646500G Supplier TCI America Supplier No. D0646500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-64-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00002497
InChI Key NEGFNJRAUMCZMY-UHFFFAOYSA-N
Synonym 3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid
PubChem CID 66837
IUPAC Name 3-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 151°C
Quantity 500 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)benzoic Acid
EINECSNumber (3)-1455
TSCA Yes