missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-2,2-dimethyl-1-propanol 97.0+%, TCI America™

Catalog No. D1727500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1727500ML 500 mL
D172725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1727500ML Supplier TCI America Supplier No. D1727500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19059-68-8
Molecular Formula C7H17NO
Molecular Weight (g/mol) 131.219
MDL Number MFCD00059879
InChI Key PYEWZDAEJUUIJX-UHFFFAOYSA-N
Synonym Dimethylaminoneopentanol
PubChem CID 87919
IUPAC Name 3-(dimethylamino)-2,2-dimethylpropan-1-ol
SMILES CC(C)(CN(C)C)CO

Specifications

Color Colorless
Boiling Point 69°C
Quantity 500 mL
UN Number 2924
Formula Weight 131.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Dimethylamino-2,2-dimethyl-1-propanol
EINECSNumber (2)-0353
TSCA Yes