accessibility menu, dialog, popup

UserName

CAS RN 19059-68-8

IUPAC Name: 3-(dimethylamino)-2,2-dimethylpropan-1-ol
Synonyms: 3-(dimethylamino)-2,2-dimethylpropan-1-ol
Molecular Weight (g/mol): 131.22
Molecular Formula: C7H17NO
InChi Key: PYEWZDAEJUUIJX-UHFFFAOYSA-N
SMILES: CN(C)CC(C)(C)CO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results