missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Dimethylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™
$115.57 - $462.29
Chemical Identifiers
| CAS | 13636-02-7 |
|---|---|
| Molecular Formula | C9H15NOS |
| Molecular Weight (g/mol) | 185.29 |
| MDL Number | MFCD09701963,MFCD07782107 |
| InChI Key | XWCNSHMHUZCRLN-UHFFFAOYNA-N |
| Synonym | 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene |
| PubChem CID | 11586409 |
| IUPAC Name | 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol |
| SMILES | CN(C)CCC(O)C1=CC=CS1 |
Chemical Identifiers
| 13636-02-7 | |
| 185.29 | |
| XWCNSHMHUZCRLN-UHFFFAOYNA-N | |
| 11586409 | |
| CN(C)CCC(O)C1=CC=CS1 |
| C9H15NOS | |
| MFCD09701963,MFCD07782107 | |
| 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene | |
| 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol |
Specifications
| 13636-02-7 | |
| White | |
| C9H15NOS | |
| 1 g | |
| XWCNSHMHUZCRLN-UHFFFAOYNA-N | |
| 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol | |
| 11586409 | |
| ≥98.0% (GC,T) | |
| 3-(Dimethylamino)-1-(2-thienyl)-1-propanol |
| 72°C | |
| 290°C | |
| MFCD09701963,MFCD07782107 | |
| 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene | |
| CN(C)CCC(O)C1=CC=CS1 | |
| 185.29 | |
| 185.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No