missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Dimethylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™
Supplier: TCI America D40095G
Specifications
| 3-(Dimethylamino)-1-(2-thienyl)-1-propanol | |
| 72°C | |
| 290°C | |
| MFCD09701963,MFCD07782107 | |
| 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene | |
| CN(C)CCC(O)C1=CC=CS1 | |
| 185.29 | |
| 185.29 | |
| Crystalline Powder |
| 13636-02-7 | |
| White | |
| C9H15NOS | |
| 5 g | |
| XWCNSHMHUZCRLN-UHFFFAOYNA-N | |
| 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol | |
| 11586409 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 13636-02-7 | |
| 185.29 | |
| XWCNSHMHUZCRLN-UHFFFAOYNA-N | |
| 11586409 | |
| CN(C)CCC(O)C1=CC=CS1 |
| C9H15NOS | |
| MFCD09701963,MFCD07782107 | |
| 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene | |
| 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol |
Safety and Handling
TSCA : No