missing translation for 'onlineSavingsMsg'
Learn More

3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane 98.0+%, TCI America™

Catalog No. D52645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52645G 5 g
D526425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52645G Supplier TCI America Supplier No. D52645G
Only null left

Chemical Identifiers

CAS 60111-47-9
Molecular Formula C18H32O3Si4
Molecular Weight (g/mol) 408.791
MDL Number MFCD00156495
InChI Key XYVYGTWMOAIWOG-UHFFFAOYSA-N
Synonym Tris[dimethyl(vinyl)siloxy]phenylsilane
PubChem CID 6453907
IUPAC Name tris[[ethenyl(dimethyl)silyl]oxy]-phenylsilane
SMILES C[Si](C)(C=C)O[Si](C1=CC=CC=C1)(O[Si](C)(C)C=C)O[Si](C)(C)C=C

Specifications

Color Colorless
Quantity 5 g
Formula Weight 408.79
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-[[Dimethyl(vinyl)silyl]oxy]-1,1,5,5-tetramethyl-3-phenyl-1,5-divinyltrisiloxane
EINECSNumber (7)-0481
TSCA No