missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[Dimethoxy(methyl)silyl]propyl Methacrylate 98.0+%, TCI America™
Supplier: TCI America M153025ML
Specifications
| 3-[Dimethoxy(methyl)silyl]propyl Methacrylate | |
| Colorless | |
| C10H20O4Si | |
| 25 mL | |
| LZMNXXQIQIHFGC-UHFFFAOYSA-N | |
| 3-[dimethoxy(methyl)silyl]propyl 2-methylprop-2-enoate | |
| 84485 | |
| ≥98.0% (GC) |
| 14513-34-9 | |
| 71°C | |
| MFCD00156475 | |
| Methacrylic Acid 3-[Dimethoxy(methyl)silyl]propyl Ester | |
| CC(=C)C(=O)OCCC[Si](C)(OC)OC | |
| 232.351 | |
| 232.35 | |
| Liquid |
Chemical Identifiers
| 14513-34-9 | |
| 232.351 | |
| LZMNXXQIQIHFGC-UHFFFAOYSA-N | |
| 84485 | |
| CC(=C)C(=O)OCCC[Si](C)(OC)OC |
| C10H20O4Si | |
| MFCD00156475 | |
| Methacrylic Acid 3-[Dimethoxy(methyl)silyl]propyl Ester | |
| 3-[dimethoxy(methyl)silyl]propyl 2-methylprop-2-enoate |
Safety and Handling
EINECSNumber : (2)-2075
TSCA : No