CAS RN 14513-34-9
CAS RN 14513-34-9
IUPAC Name:
3-[dimethoxy(methyl)silyl]propyl 2-methylprop-2-enoate
Synonyms:
3-[dimethoxy(methyl)silyl]propyl 2-methylprop-2-enoate
Molecular Weight (g/mol):
232.35
Molecular Formula:
C10H20O4Si
InChi Key:
LZMNXXQIQIHFGC-UHFFFAOYSA-N
SMILES:
CO[Si](C)(CCCOC(=O)C(C)=C)OC
1
–
1
of
1
results
1
3-[Dimethoxy(methyl)silyl]propyl Methacrylate 98.0+%, TCI America™
CAS: 14513-34-9 Molecular Formula: C10H20O4Si Molecular Weight (g/mol): 232.351 MDL Number: MFCD00156475 InChI Key: LZMNXXQIQIHFGC-UHFFFAOYSA-N Synonym: Methacrylic Acid 3-[Dimethoxy(methyl)silyl]propyl Ester PubChem CID: 84485 IUPAC Name: 3-[dimethoxy(methyl)silyl]propyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCCC[Si](C)(OC)OC