missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethoxy)benzyl alcohol

Catalog No. AAH5034503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5034503 1 g
AAH5034506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5034503 Supplier Thermo Scientific Chemicals Supplier No. H5034503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 125903-81-3 | C8H8F2O2 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 125903-81-3
Molecular Formula C8H8F2O2
Molecular Weight (g/mol) 174.15
MDL Number MFCD00236227
InChI Key BBDUCPSYPRGPGO-UHFFFAOYSA-N
Synonym 3-difluoromethoxy benzyl alcohol, 3-difluoromethoxy phenyl methanol, 3-difluoromethoxy phenyl methan-1-ol, acmc-1c79n, benzenemethanol, 3-difluoromethoxy, alpha,alpha-difluoro-3-hydroxymethyl anisole, 3-difluoromethoxy phenyl methanol
PubChem CID 2736994
IUPAC Name [3-(difluoromethoxy)phenyl]methanol
SMILES OCC1=CC(OC(F)F)=CC=C1

Specifications

Quantity 1 g
Formula Weight 174.15
Chemical Name or Material 3-(Difluoromethoxy)benzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only