missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Diethylamino)pyrrolidine 98.0+%, TCI America™
$507.02 - $507.02
Chemical Identifiers
| CAS | 20984-81-0 |
|---|---|
| Molecular Formula | C8H18N2 |
| Molecular Weight (g/mol) | 142.25 |
| MDL Number | MFCD02184351 |
| InChI Key | XHOADYKJSBCVBJ-UHFFFAOYNA-N |
| PubChem CID | 19961995 |
| IUPAC Name | N,N-diethylpyrrolidin-3-amine |
| SMILES | CCN(CC)C1CCNC1 |
Chemical Identifiers
| 20984-81-0 | |
| 142.25 | |
| XHOADYKJSBCVBJ-UHFFFAOYNA-N | |
| N,N-diethylpyrrolidin-3-amine |
| C8H18N2 | |
| MFCD02184351 | |
| 19961995 | |
| CCN(CC)C1CCNC1 |
Specifications
| 20984-81-0 | |
| 84°C | |
| MFCD02184351 | |
| 2735 | |
| CCN(CC)C1CCNC1 | |
| 142.25 | |
| 142.25 | |
| Liquid |
| Yellow | |
| C8H18N2 | |
| 5 g | |
| XHOADYKJSBCVBJ-UHFFFAOYNA-N | |
| N,N-diethylpyrrolidin-3-amine | |
| 19961995 | |
| ≥98.0% (T) | |
| 3-(Diethylamino)pyrrolidine |
Safety and Handling
TSCA : No