CAS RN 20984-81-0
CAS RN 20984-81-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(Diethylamino)pyrrolidine 98.0+%, TCI America™
CAS: 20984-81-0 Molecular Formula: C8H18N2 Molecular Weight (g/mol): 142.25 MDL Number: MFCD02184351 InChI Key: XHOADYKJSBCVBJ-UHFFFAOYNA-N PubChem CID: 19961995 IUPAC Name: N,N-diethylpyrrolidin-3-amine SMILES: CCN(CC)C1CCNC1