missing translation for 'onlineSavingsMsg'
Learn More

3-(Diethylamino)pyrrolidine 98.0+%, TCI America™

Catalog No. D18575G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D18575G Supplier TCI America Supplier No. D18575G
Only null left

Chemical Identifiers

CAS 20984-81-0
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.25
MDL Number MFCD02184351
InChI Key XHOADYKJSBCVBJ-UHFFFAOYNA-N
PubChem CID 19961995
IUPAC Name N,N-diethylpyrrolidin-3-amine
SMILES CCN(CC)C1CCNC1

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 g
UN Number 2735
Formula Weight 142.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-(Diethylamino)pyrrolidine