missing translation for 'onlineSavingsMsg'
Learn More

3-Decyn-1-ol, 97%

CAS: 51721-39-2 | C10H18O | 154.253 g/mol

$231.55 - $778.21

Chemical Identifiers

CAS 51721-39-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00040917
InChI Key YGEQBZUDPQQIFI-UHFFFAOYSA-N
Synonym 3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
PubChem CID 103940
IUPAC Name dec-3-yn-1-ol
SMILES CCCCCCC#CCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAB2083706
View Documents Promotion Details
N/A 5 g
Each for $231.55
Only null left
 
AAB2083714
View Documents Promotion Details
N/A 25 g
Each for $778.21
Only null left
 
Description

Description

3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3?-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3′-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

Solubility
Sparingly soluble in water (0.21 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

51721-39-2
154.253
YGEQBZUDPQQIFI-UHFFFAOYSA-N
103940
CCCCCCC#CCCO
C10H18O
MFCD00040917
3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
dec-3-yn-1-ol

Specifications

51721-39-2
129°C to 130°C (20 mmHg)
C10H18O
MFCD00040917
3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
YGEQBZUDPQQIFI-UHFFFAOYSA-N
dec-3-yn-1-ol
103940
97%
0.871
106°C (222°F)
1.459
1750909
Sparingly soluble in water (0.21g/L) (25°C).
CCCCCCC#CCCO
154.253
154.25
3-Decyn-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only