missing translation for 'onlineSavingsMsg'
Learn More

3-Decyn-1-ol, 97%

Catalog No. AAB2083706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2083706 5 g
AAB2083714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2083706 Supplier Thermo Scientific Chemicals Supplier No. B2083706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51721-39-2 | C10H18O | 154.253 g/mol

3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3?-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3′-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

Solubility
Sparingly soluble in water (0.21 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 51721-39-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00040917
InChI Key YGEQBZUDPQQIFI-UHFFFAOYSA-N
Synonym 3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
PubChem CID 103940
IUPAC Name dec-3-yn-1-ol
SMILES CCCCCCC#CCCO

Specifications

Density 0.871
Boiling Point 129°C to 130°C (20 mmHg)
Flash Point 106°C (222°F)
Refractive Index 1.459
Quantity 5 g
Beilstein 1750909
Solubility Information Sparingly soluble in water (0.21g/L) (25°C).
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material 3-Decyn-1-ol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only