missing translation for 'onlineSavingsMsg'
Learn More

3-Decyn-1-ol, 97%

CAS: 51721-39-2 | C10H18O | 154.253 g/mol

Supplier:  Thermo Scientific Chemicals B2083714

 View more versions of this product

Catalog No. AAB2083714


Only null left

Description

Description

3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3?-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3′-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

Solubility
Sparingly soluble in water (0.21 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Decyn-1-ol
0.871
106°C (222°F)
1.459
25 g
3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
YGEQBZUDPQQIFI-UHFFFAOYSA-N
dec-3-yn-1-ol
103940
97%
51721-39-2
129°C to 130°C (20 mmHg)
C10H18O
MFCD00040917
1750909
Sparingly soluble in water (0.21g/L) (25°C).
CCCCCCC#CCCO
154.253
154.25

Chemical Identifiers

51721-39-2
154.253
YGEQBZUDPQQIFI-UHFFFAOYSA-N
103940
CCCCCCC#CCCO
C10H18O
MFCD00040917
3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
dec-3-yn-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only