missing translation for 'onlineSavingsMsg'
Learn More

3-Decen-2-one 93.0+%, TCI America™

Catalog No. p-7127022
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19335ML 5 mL
D193325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19335ML Supplier TCI America Supplier No. D19335ML
Only null left

Chemical Identifiers

CAS 10519-33-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00015700
InChI Key JRPDANVNRUIUAB-CMDGGOBGSA-N
Synonym 3-decen-2-one, heptylidene acetone, oenanthylidene acetone, e-3-decen-2-one, unii-z22804bqxd, 3e-dec-3-en-2-one, e-dec-3-en-2-one, dec-3-en-2-one, fema no. 3532, enanthylidene acetone
PubChem CID 5363233
IUPAC Name (E)-dec-3-en-2-one
SMILES CCCCCCC=CC(=O)C

Specifications

Color Yellow
Boiling Point 109°C
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Decen-2-one