missing translation for 'onlineSavingsMsg'
Learn More

3-Decanol, 97%

Catalog No. AAB2414114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2414114 25 g
AAB2414106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2414114 Supplier Thermo Scientific Chemicals Supplier No. B2414114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1565-81-7 | C10H22O | 158.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1565-81-7
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00021956
InChI Key ICEQLCZWZXUUIJ-UHFFFAOYSA-N
Synonym 3-decanol, heptyl ethyl carbinol, ethyl heptyl carbinol, 3-decanol, r, 3-decanol, s, fema no. 3605, 3-decyl alcohol, ethylheptylcarbinol, deca-8-yl alcohol, acmc-20mxdt
PubChem CID 519158
IUPAC Name decan-3-ol
SMILES CCCCCCCC(CC)O

Specifications

Density 0.83
Boiling Point 213°C
Flash Point 99°C (210°F)
Refractive Index 1.435
Quantity 25 g
Beilstein 1719658
Formula Weight 158.29
Percent Purity 97%
Chemical Name or Material 3-Decanol
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only