CAS RN 1565-81-7
IUPAC Name:
decan-3-ol
Synonyms:
decan-3-ol
Molecular Weight (g/mol):
158.29
Molecular Formula:
C10H22O
InChi Key:
ICEQLCZWZXUUIJ-UHFFFAOYNA-N
SMILES:
CCCCCCCC(O)CC
1
–
1
of
1
results
1
3-Decanol, 97%
CAS: 1565-81-7 Molecular Formula: C10H22O Molecular Weight (g/mol): 158.285 MDL Number: MFCD00021956 InChI Key: ICEQLCZWZXUUIJ-UHFFFAOYSA-N Synonym: 3-decanol,heptyl ethyl carbinol,ethyl heptyl carbinol,3-decanol, r,3-decanol, s,fema no. 3605,3-decyl alcohol,ethylheptylcarbinol,deca-8-yl alcohol,acmc-20mxdt PubChem CID: 519158 IUPAC Name: decan-3-ol SMILES: CCCCCCCC(CC)O