missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid, 98%

Catalog No. p-7055498
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6409803 1 g
AAH6409806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6409803 Supplier Thermo Scientific Chemicals Supplier No. H6409803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 162401-62-9 | C12H12F2O4 | 258.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162401-62-9
Molecular Formula C12H12F2O4
Molecular Weight (g/mol) 258.221
MDL Number MFCD04621687
InChI Key IGFDIFLMMLWKKY-UHFFFAOYSA-N
Synonym 3-cyclopropylmethoxy-4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy, roflumilast intermediate, roflumilast intermediate b, 3-cyclopropylmethoxy-4-difluoromethoxybenzoicacid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, 3-cyclopropylmethoxy 4-difluoromethoxy benzoic acid
PubChem CID 2394006
IUPAC Name 3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
SMILES C1CC1COC2=C(C=CC(=C2)C(=O)O)OC(F)F

Specifications

Melting Point 118°C to 120°C
Quantity 1 g
Beilstein 11413145
Formula Weight 258.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 605-288-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only