missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropyl-1H-pyrazole, 98%

Catalog No. p-7055479
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6406903 1 g
AAH6406906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6406903 Supplier Thermo Scientific Chemicals Supplier No. H6406903

CAS: 100114-57-6 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100114-57-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD04970938,MFCD06739298
InChI Key TXWDVWSJMDFNQY-UHFFFAOYSA-N
PubChem CID 7017816
IUPAC Name 5-cyclopropyl-1H-pyrazole
SMILES C1CC1C1=CC=NN1

Specifications

Color Colorless
Quantity 1 g
Beilstein 1561659
Synonym 3-cyclopropyl-1h-pyrazole, 5-cyclopropylpyrazole, 1h-pyrazole, 3-cyclopropyl, 3-cyclopropylpyrazole, 1h-pyrazole,3-cyclopropyl, 3-cyclopropyl-2h-pyrazole, 3-cyclopropyl-1h-pyrazole 97+%, acmc-2097ms, 1h-pyrazole, 5-cyclopropyl
Formula Weight 108.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Cyclopropyl-1H-pyrazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only