accessibility menu, dialog, popup

UserName

NNH

CAS RN 100114-57-6

IUPAC Name: 5-cyclopropyl-1H-pyrazole
Synonyms: 3-cyclopropyl-2H-pyrazole
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: TXWDVWSJMDFNQY-UHFFFAOYSA-N
SMILES: C1CC1C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results