missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyclopentylpropionic Acid 98.0+%, TCI America™
Supplier: TCI America C293425G
Specifications
| 3-Cyclopentylpropionic Acid | |
| Yellow | |
| C8H14O2 | |
| 25 g | |
| 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate | |
| OC(=O)CCC1CCCC1 | |
| 142.20 | |
| CHEBI:50899 | |
| ≥98.0% (GC,T) |
| 140-77-2 | |
| 123°C | |
| MFCD00001392 | |
| 1993 | |
| ZRPLANDPDWYOMZ-UHFFFAOYSA-N | |
| 3-cyclopentylpropanoic acid | |
| 8818 | |
| 142.20 | |
| Liquid |
Chemical Identifiers
| 140-77-2 | |
| 142.20 | |
| ZRPLANDPDWYOMZ-UHFFFAOYSA-N | |
| 8818 | |
| 3-cyclopentylpropanoic acid |
| C8H14O2 | |
| MFCD00001392 | |
| 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate | |
| CHEBI:50899 | |
| OC(=O)CCC1CCCC1 |
Safety and Handling
TSCA : Yes