missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionic Acid 98.0+%, TCI America™

Catalog No. p-7125731
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29345G 5 g
C293425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29345G Supplier TCI America Supplier No. C29345G
Only null left

Chemical Identifiers

CAS 140-77-2
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001392
InChI Key ZRPLANDPDWYOMZ-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
PubChem CID 8818
ChEBI CHEBI:50899
IUPAC Name 3-cyclopentylpropanoic acid
SMILES OC(=O)CCC1CCCC1

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
UN Number 1993
Formula Weight 142.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Cyclopentylpropionic Acid