missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionic acid, 98%

CAS: 140-77-2 | C8H14O2 | 142.20 g/mol

Supplier:  Thermo Scientific Chemicals 111610250

 View more versions of this product

Catalog No. AC111610250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Cyclopentylpropionic acid
140-77-2
100.0
0.9900g/mL
46°C
98%
C8H14O2
C5H9CH2CH2CO2H
25 g
3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
ZRPLANDPDWYOMZ-UHFFFAOYSA-N
3-cyclopentylpropanoic acid
8818
142.2
Liquid
98%
97.5
Colorless to Yellow
130.0°C to 132.0°C (12.0 mmHg)
Authentic
Glass bottle
1.4560 to 1.4580
MFCD00001392
0.99
Solubility in water: insoluble
OC(=O)CCC1CCCC1
142.20
CHEBI:50899
98%

Chemical Identifiers

140-77-2
142.20
ZRPLANDPDWYOMZ-UHFFFAOYSA-N
8818
3-cyclopentylpropanoic acid
C8H14O2
MFCD00001392
3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
CHEBI:50899
OC(=O)CCC1CCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-433-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only