missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionic acid, 98%

Catalog No. p-97192
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111610050 5 g Glass bottle
AC111610250 25 g Glass bottle
AC111611000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111610050 Supplier Thermo Scientific Chemicals Supplier No. 111610050

CAS: 140-77-2 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-77-2
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001392
InChI Key ZRPLANDPDWYOMZ-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
PubChem CID 8818
ChEBI CHEBI:50899
IUPAC Name 3-cyclopentylpropanoic acid
SMILES OC(=O)CCC1CCCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 130.0°C to 132.0°C (12.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4560 to 1.4580
Linear Formula C5H9CH2CH2CO2H
Quantity 5 g
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble
Formula Weight 142.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Cyclopentylpropionic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-433-0
TSCA TSCA

RUO – Research Use Only